missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562063
This item is not returnable.
View return policy
Description
Immunogen sequence: MVDFDEGSDV FQMLNMNSAP TFINFP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
MAGT1 encodes a magnesium cation transporter protein that localizes to the cell membrane. This protein also associates with N-oligosaccharyl transferase and therefore may have a role in N-glycosylation. Mutations in this gene cause mental retardation X-linked type 95 (MRX95). This gene may have multiple in-frame translation initiation sites, one of which would encode a shorter protein with an N-terminus containing a signal peptide at amino acids 1-29.
Specifications
| MAGT1 | |
| Polyclonal | |
| Unconjugated | |
| Magt1 | |
| 2410001C15Rik; 2610529C04Rik; 2810482I07Rik; bA217H1.1; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit MAGT1; Iag2; IAP; Implantation-associated protein; magnesium transporter 1; magnesium transporter protein 1; MAGT1; MRX95; Oligosaccharyl transferase subunit MAGT1; oligosaccharyltransferase 3 homolog B; OST3B; PRO0756; PSEC0084; UNQ628/PRO1244; XMEN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84061 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0U3 | |
| Magt1 | |
| Recombinant protein corresponding to Human MAGT1. Recombinant protein control fragment (Product #RP-99623). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction