Learn More
Invitrogen™ MAGPIX™ Drive Fluid Plus
Product Code. 30245082
Change view
Click to view available options
Quantity:
4 x 700 mL
Unit Size:
Pack of 4
Product Code. Quantity Unit Size
30245082 4 x 700 mL Pack of 4
1 options
Product Code. 30245082 Supplier Invitrogen™ Supplier No. 4050030
  • ONLINE EXCLUSIVE PRICE
    £73.00
    £68.65 / Pack of 4
    Save £4.35
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
MAGPIX Drive Fluid Plus is the delivery medium for transporting the sample being analyzed on the Luminex MAGPIX system to the optic components.
MAGPIX Drive Fluid Plus is the delivery medium for transporting the sample being analyzed on the Luminex MAGPIX system to the optic components. Each unit in the 4-pack of drive fluid contains 700 mL.
Please refer to the Luminex instrument user manual for proper installation and use of MAGPIX Drive Fluid Plus with the MAGPIX System.
This reformulated sheath product replaces the original MAGPIX Drive Fluid (Cat. No. MPXDF4PK). The new sheath formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.
For Research Use Only. Not for use in diagnostic procedures. Not intended for human or animal therapeutic or diagnostic use
TRUSTED_SUSTAINABILITY
Specifications
Product Line MAGPIX
Quantity 4 x 700 mL