Learn More
Invitrogen™ MAGOH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596493
Description
Immunogen sequence: MESDFYLRYY VGHKGKFGHE FLEFEFRPDG KLRYANNSNY KNDVMIRKEA YVHKSVMEEL KRIIDDSEIT KEDDALWPPP DRVGRQELEI VIGDEHISFT TSKIGSLIDV NQSKDPEGLR VFYYLVQDLK CLVFSLIGLH FKIKPI; Positive Samples: SKOV3, HeLa, MCF7, Jurkat, Mouse spleen, Mouse thymus, Rat spleen; Cellular Location: Cytoplasm, Nucleus, Nucleus speckle.
Drosophila that have mutations in their mago nashi gene produce progeny with defects in germplasm assembly and germline development. This gene encodes the mammalian mago nashi homolog. In mammals, mRNA expression is not limited to the germ plasm, but is expressed ubiquitously in adult tissues and can be induced by serum stimulation of quiescent fibroblasts.
Specifications
| MAGOH | |
| Polyclonal | |
| Unconjugated | |
| MAGOH | |
| ch211-216k22.5; hypothetical protein LOC550395; mago homolog, exon junction complex core component; mago homolog, exon junction complex subunit; mago nashi; Magoh; MAGOH1; MAGOHA; Mago-m; mago-nashi homolog, proliferation-associated; mago-nashi homolog, proliferation-associated (Drosophila); mago-nashi-like proliferation-associated protein; Mos2; Protein mago nashi homolog; RP5-1024G6.4; zgc:112220 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17149, 298385, 4116 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.08 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61326, P61327, Q27W02 | |
| MAGOH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-146 of human MAGOH (NP_0023611). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.