missing translation for 'onlineSavingsMsg'
Learn More

Magnolin, MedChemExpress

Product Code. 16463916
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463916 25 mg 25mg
16451628 5 mg 5mg
16419218 10 mg 10mg
16404917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16463916 Supplier Medchem Express Supplier No. HYN1374
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC50s of 87 nM and 16.5 nM, respectively.

Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC50s of 87 nM and 16.5 nM, respectively.

Specifications

Chemical Name or Material Magnolin
CAS 31008-18-1
Color White
Purity Grade Notes Research
Molecular Formula C23H28O7
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (240.12 mM; Need ultrasonic)
SMILES COC1=C(OC)C(OC)=CC([C@H]2OC[C@]3([H])[C@@H](C4=CC=C(OC)C(OC)=C4)OC[C@@]32[H])=C1
Molecular Weight (g/mol) 416.46
Formula Weight 416.46
Percent Purity 98.85%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.