missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Streptavidin Negative Selection Beads

Product Code. 15572567
Click to view available options
Quantity:
200 Tests
25 Tests
Unit Size:
200 tests
25 tests
This item is not returnable. View return policy
Contains Sodium azide

Product Code. 15572567

Brand: Invitrogen™ MSNB600274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Sodium azide

Designed for the magnetic separation of cells by negative selection. Undesired cells are bound by a cocktail of biotinylated antibodies and then streptavidin coated magnetic beads.

  • Clone and concentration of biotinylated antibody for undesired cell labeling, as well as concentration of MagniSort™ Streptavidin Negative Selection Beads must be determined empirically
  • Please refer to the protocol for details on how to optimize both the antibody or antibody cocktail and MagniSort™ Streptavidin Negative Selection Beads.
  • For depletion of cells using a single antibody, we recoomend the MagniSort™ Streptavidin Positive Selection Beads (cat. MSPB-6003) and protocol
TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 Tests
Form Liquid
Ligand Streptavidin
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin

For Research Use Only. Not for use in diagnostic procedures.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado