Learn More
Invitrogen™ MagniSort™ Mouse F4/80 Positive Selection Kit
Designed for the magnetic separation of F4/80+ cells by positive selection. It has been optimized for the isolation of F4/80+ cells from mouse bone marrow or splenocytes utilizing a biotinylated Anti-Mouse F4/80 antibody and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8802-6863-74
Code : NEW
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Includes: MagniSort™ Anti-Mouse F4/80 Biotin : 200 tests, 20μL/test
MagniSort™ Positive Selection Beads B
Description
- F4/80+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from F4/80+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse F4/80, clone BM8 and Anti-Mouse CD11b, clones M1/70
- To reduce non-specific binding, the MagniSort™ Anti-Mouse F4/80 Biotin contains Fc Receptor block
Specifications
F4/80+ | |
Positive Selection Kit | |
Mouse |
For Research Use Only. Not for use in diagnostic procedures.