missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD8 T cell Enrichment Kit

Product Code. 15581277
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
This item is not returnable. View return policy
Contains Sodium azide

Product Code. 15581277

Brand: Invitrogen™ 8804682274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Sodium azide

Includes

  • MagniSort Mouse CD8 T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic separation of mouse CD8 T cells by negative selection. It has been optimized for the isolation of mouse CD8 T cells from spleens or lymph nodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 T cells untouched and free in solution.

The MagniSort™ Mouse CD8 T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD4 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD24 Biotin
Anti-Mouse CD45R (B220) Biotin
Anti-Mouse CD49b (Integrin alpha 2) Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse γδ TCR Biotin
Anti-Mouse TER-119 Biotin
TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (CD8)
Product Type Cell Enrichment Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.