Learn More
Invitrogen™ MagniSort™ Mouse CD8 Positive Selection Kit
Designed for the magnetic separation of CD8+ cells by positive selection. It has been optimized for the isolation of CD8+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD8a antibody and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8802-6842-74
Code : NEW
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Includes: MagniSort Anti-Mouse CD8 Biotin : 200 tests, 20μL/test
MagniSort Positive Selection Beads B
Description
- CD8+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from CD8+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD8a, clone 53-6.7
Specifications
CD8+ | |
Positive Selection Kit | |
Mouse |
For Research Use Only. Not for use in diagnostic procedures.