missing translation for 'onlineSavingsMsg'
Learn More

eBioscience™ MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit

Product Code. 15501287
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15501287 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains Sodium azide
Product Code. 15501287 Supplier eBioscience Supplier No. 8804682574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Sodium azide

Includes

  • MagniSort Mouse CD8 Naïve T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic separation of CD8 naïve T cells by negative selection. It has been optimized for the isolation of mouse CD8 naïve T cells from spleens or lymphnodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 naïve T cells untouched and free in solution.

The MagniSort™ Mouse CD8 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD4 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD24 Biotin
Anti-Mouse CD44 Biotin
Anti-Mouse CD45R (B220) Biotin
Anti-Mouse CD49b (Integrin alpha 2) Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse γδ TCR Biotin
Anti-Mouse TER-119 Biotin
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.