missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD49b Positive Selection Kit

Product Code. 15521267
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
This item is not returnable. View return policy
Contains Sodium azide

Product Code. 15521267

Brand: Invitrogen™ 8802686274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Sodium azide

Includes

  • MagniSort™ Anti-Mouse CD49b Biotin : 200 tests, 20μL/test
  • MagniSort™ Positive Selection Beads A

Designed for the magnetic separation of CD49b+ cells by positive selection. It has been optimized for the isolation of CD49b+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD49b antibody and streptavidin-coated magnetic beads.

  • CD49b+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD49b+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD49b (integrin alpha 2), clone DX5, and Anti-Mouse NK1.1, clone PK136. For mouse strains that do not express NK1.1 antigen such as BALB/c, SJL, AKR, CBA C3H, anti-Mouse CD335 (NKp46), clone 29A1.4, may be used
  • For higher purity of CD49b+ cells, 10 μL/test of beads may be used, instead of 20 μL as indicated in the protocol, however, this may result in decreased recovery rate of CD49b+ cells
TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD49b+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.