missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagniSort™ Mouse CD45 Positive Selection Kit
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
Description
Includes
- MagniSort Anti-Mouse CD45 Biotin B : 200 tests, 20μL/test
- MagniSort Positive Selection Beads A
- CD45+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from CD45+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD45, clone 30-F11
Specifications
Specifications
| Cell Type | CD45+ |
| Product Type | Positive Selection Kit |
| No. of Tests | 200 tests |
| Quantity | 200 tests |
| Target Species | Mouse |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction