missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD45.1 Positive Selection Kit

Product Code. 15581257
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
This item is not returnable. View return policy
Contains Sodium azide

Product Code. 15581257

Brand: Invitrogen™ 8802684874

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Sodium azide

Includes

  • MagniSort™ Anti-Mouse CD45.1 Biotin : 200 tests, 20μL/test
  • MagniSort™ Positive Selection Beads A

Designed for the magnetic separation of CD45.1+ cells by positive selection. It has been optimized for the isolation of CD45.1+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD45.1 antibody and streptavidin-coated magnetic beads.

  • CD45.1+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD45.1+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD45.1, clone A20
  • When the starting frequency of CD45.1 cells is greater than 75% of the total cells, the amount of MagniSort™ Magnetic Positive Selection Beads to be used can be increased to 40 μL/test to maximize recovery
TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD45.1+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.