missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagniSort™ Mouse CD11c Positive Selection Kit
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
Description
Includes
- MagniSort™ Mouse CD11c Biotin : 200 tests, 20μL/test
- MagniSort™ Positive Selection Beads B
- CD11c+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from CD11c+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD11c, clone N418 and Anti-Mouse CD11b, clone M1/70
- To reduce non-specific binding, the MagniSort™ Anti-Mouse CD11c Biotin contains Fc Receptor block
Specifications
Specifications
| Cell Type | CD11c+ |
| Product Type | Positive Selection Kit |
| No. of Tests | 200 tests |
| Quantity | 200 tests |
| Target Species | Mouse |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction