Learn More
Invitrogen™ MagniSort™ Mouse CD117 (c-Kit) Positive Selection Kit
Designed for the magnetic separation of CD117+ cells by positive selection. It has been optimized for the isolation of CD117+ cells from mouse bone marrow utilizing a biotinylated Anti-Mouse CD117 antibody and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8802-6838-74
Code : NEW
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Includes: MagniSort Anti-Mouse CD117 (c-Kit) Biotin : 200 tests, 20μL/test
MagniSort Positive Selection Beads B
Description
- CD117+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from CD117+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD117, clone 2B8
Specifications
CD117+ | |
Positive Selection Kit | |
Mouse |
For Research Use Only. Not for use in diagnostic procedures.