missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagniSort™ Mouse B cell Enrichment Kit
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
Description
Includes
- MagniSort Mouse B cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
- MagniSort Negative Selection Beads B
Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave B cells untouched and free in solution.
The MagniSort™ Mouse B cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD3 Biotin
Anti-Mouse CD4 Biotin
Anti-Mouse CD8 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD49b Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse TER-119 Biotin
The MagniSort™ Mouse B cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD3 Biotin
Anti-Mouse CD4 Biotin
Anti-Mouse CD8 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD49b Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse TER-119 Biotin
Specifications
Specifications
| Cell Type | B Cells |
| Product Type | Cell Enrichment Kit |
| No. of Tests | 200 tests |
| Quantity | 200 tests |
| Target Species | Mouse |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction