missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ MagniSort™ Human CD8 Memory T cell Enrichment Kit

Product Code. 15521277
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
This item is not returnable. View return policy
Contains Sodium azide

Product Code. 15521277

Brand: Invitrogen™ 8804681574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Sodium azide

Includes

  • MagniSort Human CD8 Memory T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads A

Designed for the magnetic separation of CD8 memory T cells by negative selection. It has been optimized for the isolation of human CD8 memory T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 memory T cells untouched and free in solution.

The MagniSort™ Human CD8 Memory T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD4 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RA Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.