Learn More
Invitrogen™ MagniSort™ Human CD45 Depletion Kit
Designed to deplete CD45+ cells from human lysed whole blood or peripheral blood mononuclear cells, utilizing a biotinylated Anti-Human CD45 antibody and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8804-6802-74
Code : NEW
Additional Details : Weight : 0.23750kg
Alert:
Contains Sodium azide
Includes: MagniSort Human CD45 Biotin A : 200 tests, 20μL/test
MagniSort Negative Selection Beads B
Description
- CD45+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the CD45- cells can be separated from CD45+ cells by decanting
- After depletion, the purity of selected cells can be verified by staining with Anti-Human CD45, clone 2D1
Specifications
CD45+ | |
Cell Depletion Kit | |
Human |
For Research Use Only. Not for use in diagnostic procedures.