missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagniSort™ Human CD4 Naïve T cell Enrichment Kit
Description
Includes
- MagniSort Human CD4 Naïve T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
- MagniSort Negative Selection Beads A
Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 naïve T cells untouched and free in solution.
The MagniSort™ Human CD4 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD8 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RO Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin
The MagniSort™ Human CD4 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD8 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RO Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin
Specifications
Specifications
| Cell Type | T Cells (Naïve, CD4) |
| Product Type | Cell Enrichment Kit |
| No. of Tests | 200 tests |
| Quantity | 200 tests |
| Target Species | Human |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?