missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Magnetic Plastic Radiation Transport Sign

Product Code. 11770983
Change view
Click to view available options
Displayed Text:
Radioactive material 7A I
Radioactive substances
Radioactive substances 7B II
Radioactive substances 7C III
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text unitSize
11770983 Radioactive material 7A I Each
11720254 Radioactive substances Each
11710993 Radioactive substances 7B II Each
11760993 Radioactive substances 7C III Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11770983 Supplier Brady™ Supplier No. 811664

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance Class 7: Radioactive material
Material Plastic
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Magnetic Plastic
For Use With (Application) Stickers for the transport of dangerous goods are eye-catching and proven in practice
Height (Metric) 297 mm
Width (Metric) 297 mm
Shape Diamond
Languages English
Displayed Text Radioactive material 7A I
Product Type Radioactive Transport Sign
Color Black on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.