Learn More
Specac™ Magnetic Film Holder
Product Code. 15269997
Encompass_Preferred
Click to view available options
Type:
Magnetic Film Holders
Unit Size:
Each
Product Code. 15269997
Brand: Specac™ GS03820
Magnetic Film Holders are designed to hold a thin film post Production for direct placement into an FTIR instrument for analysis.
Specifications
Type Magnetic Film Holders