Learn More
Manutan Magnetic easy wipe racking strip, green, 10m x 90mm
Product Code. 12806078
Encompass_Preferred
Product Code. 12806078 Supplier Manutan Supplier No. A338084
Only null left
Add to basket
Add to basket
Cut with scissors to provide an easy, flexible solution to location identification. 10m long plasticised magnetic marker strips. Write on and wipe off using standard wet wipe marker pens. Ideal for cold stores.
Roll width : 90mm