missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Magnetic easy wipe racking strip, green, 10m x 40mm
Brand: Manutan A338135
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Cut with scissors to provide an easy, flexible solution to location identification. 10m long plasticised magnetic marker strips. Write on and wipe off using standard wet wipe marker pens. Ideal for cold stores.Roll width : 40mm