Learn More
Manutan Magnetic easy wipe racking strip, green, 10m x 10mm
Product Code. 12851639
Encompass_Preferred
Product Code. 12851639 Supplier Manutan Supplier No. A338140
Only null left
Add to basket
Add to basket
Cut with scissors to provide an easy, flexible solution to location identification. 10m long plasticised magnetic marker strips. Write on and wipe off using standard wet wipe marker pens. Ideal for cold stores.
Roll width : 10mm