Learn More
Manutan Magnetic easy wipe racking strip, blue, 10m x 60mm
Product Code. 12811739
Encompass_Preferred
Product Code. 12811739 Supplier Manutan Supplier No. A338152
Only null left
Add to basket
Add to basket
Cut with scissors to provide an easy, flexible solution to location identification. 10m long plasticised magnetic marker strips. Write on and wipe off using standard wet wipe marker pens. Ideal for cold stores.
Roll width : 60mm