missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Magnetic easy wipe location labels, 80mm x 20mm
Brand: Manutan A339427
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Individual magnetic labels with cleanable white surface, write on and wipe off, fix and relocate. Use with wet wipe marker pens.Dimensions (L x W): 80mm x 20mm