missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Magnetic beadsTM Streptavidin
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
The streptavidin magnetic beads are uniform size, narrow size distribution with large surface area and unique surface coating, it is easy to capture the biotinylated proteins or other molecules, and the bounded protein have no activity lost, can help us get the best performance and highly reproducible results.
Specifications
Specifications
| Kit Contents | Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L, the Biotinylated protein can bind to the SA beads irreversibly. The Magenetic beads are pre-coupled with Streptavidin tetramer |
| Content And Storage | -20°C, Avoid light |
| Shipping Condition | RT |
| Quantity | 5 mg |
| Type | Magnetic Affinity Bead |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction