missing translation for 'onlineSavingsMsg'
Learn More

Magnesium, turnings, 99+%, Thermo Scientific Chemicals

Product Code. 11466750
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466750 2500 g 2500g
11476750 100 g 100g
11347995 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11466750 Supplier Thermo Scientific Alfa Aesar Supplier No. L08120.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Magnesium, turnings is used in solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units. It is a component in die-casting, to remove sulfur in the production of iron and steel, the production of titanium in the Kroll process. It is widely used for manufacturing of mobile phones, laptop and tablet computers, cameras, and other electronic components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium, turnings is used in solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units. It is a component in die-casting, to remove sulfur in the production of iron and steel, the production of titanium in the Kroll process. It is widely used for manufacturing of mobile phones, laptop and tablet computers, cameras, and other electronic components.

Solubility
Reacts with water.

Notes
Store away from air, moisture and oxidizing agents. Incompatible with acids, air, nitrogen and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Melting Point 650°C
Boiling Point 1091°CC
Density 1.738
Quantity 2500 g
Odor Odorless
Sensitivity Moisture sensitive
UN Number UN1869
Beilstein 4948473
Solubility Information Reacts with water.
Formula Weight 24.305
Percent Purity ≥99%
Chemical Name or Material Magnesium, turnings
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium turnings
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Self-heating substance Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H228-Flammable solid.
  • H251-Self-heating: may catch fire.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P420-Store away from other materials.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.