missing translation for 'onlineSavingsMsg'
Learn More

Magnesium tungsten oxide, 99% (metals basis), Thermo Scientific™

Product Code. 11321559
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-11-0
Molecular Formula MgO4W
Molecular Weight (g/mol) 272.141
MDL Number MFCD00016209
InChI Key DJZHPOJZOWHJPP-UHFFFAOYSA-N
Synonym magnesium tungstate, magnesium tungstate vi, magnesium tungsten oxide mgwo4, magnesium dioxido dioxo tungsten, magnesiumwolframat, magnesium wolframate, of a kind used as a luminophore, mg.wo4, tungstate wo42-, magnesium 1:1 , t-4, magnesium 2+ tungstate, magnesium tungstate-w
PubChem CID 3084161
IUPAC Name magnesium;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[Mg+2]

Specifications

Physical Form Powder
Quantity 100 g
Formula Weight 272.16
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Magnesium tungsten oxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'