missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium titanium oxide, 99% (metals basis), Thermo Scientific Chemicals
£32.40 - £94.50
Chemical Identifiers
CAS | 12032-30-3 |
---|---|
Molecular Formula | MgO3Ti |
Molecular Weight (g/mol) | 120.169 |
MDL Number | MFCD00016198 |
InChI Key | XSETZKVZGUWPFM-UHFFFAOYSA-N |
Synonym | magnesium titanate, magnesium dititanate, karrooite, ticon 75, magnesium titanium trioxide, atd 16, magnesium titanate iv mgtio3, titanium magnesium oxide timgo3, magnesium dititanium pentoxide, magnesium titanate mgti2o5 |
PubChem CID | 159407 |
IUPAC Name | magnesium;oxygen(2-);titanium(4+) |
SMILES | [O-2].[O-2].[O-2].[Mg+2].[Ti+4] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11331199
|
Thermo Scientific Alfa Aesar
011398.30 |
250 g |
£32.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321199
|
Thermo Scientific Alfa Aesar
011398.A1 |
1 kg |
£94.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Magnesium titanium oxide is used in the primary chemical intermediates and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12032-30-3 | |
120.169 | |
XSETZKVZGUWPFM-UHFFFAOYSA-N | |
159407 | |
[O-2].[O-2].[O-2].[Mg+2].[Ti+4] |
MgO3Ti | |
MFCD00016198 | |
magnesium titanate, magnesium dititanate, karrooite, ticon 75, magnesium titanium trioxide, atd 16, magnesium titanate iv mgtio3, titanium magnesium oxide timgo3, magnesium dititanium pentoxide, magnesium titanate mgti2o5 | |
magnesium;oxygen(2-);titanium(4+) |
Specifications
1610°C | |
White | |
MgO3Ti | |
magnesium titanate, magnesium dititanate, karrooite, ticon 75, magnesium titanium trioxide, atd 16, magnesium titanate iv mgtio3, titanium magnesium oxide timgo3, magnesium dititanium pentoxide, magnesium titanate mgti2o5 | |
XSETZKVZGUWPFM-UHFFFAOYSA-N | |
magnesium;oxygen(2-);titanium(4+) | |
159407 | |
99% | |
3.36 g/mL | |
Magnesium titanium oxide |
12032-30-3 | |
Powder | |
MFCD00016198 | |
Insoluble in water. | |
[O-2].[O-2].[O-2].[Mg+2].[Ti+4] | |
120.169 | |
120.21 | |
Odorless | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 234-766-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only