missing translation for 'onlineSavingsMsg'
Learn More

Magnesium thiosulfate hexahydrate, 99%

Product Code. 11498620
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11498620

Brand: Thermo Scientific Alfa Aesar 046114.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-30-5 | H12MgO9S2 | 244.512 g/mol

Magnesium hyposulfite finds application in the treatment of certain skin diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-30-5
Molecular Formula H12MgO9S2
Molecular Weight (g/mol) 244.512
MDL Number MFCD00169871
InChI Key CQDMJJVHDPDPHO-UHFFFAOYSA-L
Synonym magnesium thiosulfate hexahydrate, unii-59481j5eha, thiosulfuric acid h2s2o3 , magnesium salt 1:1 , hexahydrate 9ci, magnesium 2+ hypo hexahydrate, magnesium thiosulfate hexahydrate 100g, magnesium 2+ ion hexahydrate thiosulphate
PubChem CID 26002
IUPAC Name magnesium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=S)[O-].[Mg+2]

Specifications

Melting Point 170°C (in -3H2O)
Color White
Physical Form Crystalline
Quantity 500 g
Linear Formula MgS2O3·6H2O
Merck Index 14,5694
Solubility Information Soluble in water.
Formula Weight 244.53 (136.44 Anhydrous)
Percent Purity 99%
Density 1.818 g/mL
Chemical Name or Material Magnesium thiosulfate hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium thiosulfate hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Magnesium thiosulfate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.