missing translation for 'onlineSavingsMsg'
Learn More

Magnesium tert-Butoxide, TRC

Product Code. 30483900
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483900 1 g 1g
30477740 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483900 Supplier Toronto Research Chemicals Supplier No. TRCM1103101G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium tert-Butoxide
CAS 32149-57-8
Molecular Formula 2(C4H10O) . Mg
Quantity 1 g
Synonym tert-Butyl Alcohol, Magnesium Salt, Di(tert-butoxy)magnesium
InChI Formula InChI=1S/2C4H9O.Mg/c2*1-4(2,3)5;/h2*1-3H3;/q2*-1;+2
SMILES [O-]C(C)(C)C.[O-]C(C)(C)C.[Mg+2]
IUPAC Name magnesium 2-methylpropan-2-olate
Molecular Weight (g/mol) 172.54
Formula Weight 170.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.