missing translation for 'onlineSavingsMsg'
Learn More

Magnesium tert-butoxide, 93+%

Product Code. 10135053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10135053 50 g Plastic bottle 50g
10055553 10 g Glass Bottle 10g
10064343 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10135053 Supplier Thermo Scientific Acros Supplier No. 390470500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 32149-57-8 | C8H18MgO2 | 170.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32149-57-8
Molecular Formula C8H18MgO2
Molecular Weight (g/mol) 170.53
MDL Number MFCD03788761
InChI Key RTKCPZYOLXPARI-UHFFFAOYSA-N
Synonym magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate
PubChem CID 12040442
IUPAC Name magnesium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]

Specifications

Color White to Gray
Assay Percent Range 93+%
Packaging Plastic bottle
Quantity 50 g
Solubility Information Solubility in water: reacts violently
Formula Weight 170.53
Percent Purity 93+%
Physical Form Powder
Chemical Name or Material Magnesium tert-butoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.