missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, TRC

Product Code. 30482068
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482068 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482068 Supplier Toronto Research Chemicals Supplier No. TRCM19813050G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Sulfate
CAS 7487-88-9
Quantity 50 g
Molecular Formula MgSO4
Synonym Sulfuric acid magnesium salt (1:1) (8CI, 9CI, ACI), Magnesium sulfate (1:1) (ACI), Arrosalt 2327, Epso Top, Epsom salt, Epsom salts, M-HOS, Magnesii sulfas, Magnesium sulfate, Magnesium sulphate, MG 3K, MG-N, Mg-OK, MG-OK 100, MN 00, MN 30, MN 70, NP-YW 1, OT-S, OT-S (drying agent), SN 00, SSN 00, Sulfuric acid magnesium salt, Sulfuric acid, magnesium salt (1:1), Sunmate, TC-P, Tomix OT, USN 00, WS 1S
InChI Formula InChI=1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
SMILES [Mg+2].[O-]S(=O)(=O)[O-]
IUPAC Name magnesium;sulfate
Molecular Weight (g/mol) 120.37
Formula Weight 119.9368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.