missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Heptahydrate, TRC

Product Code. 30009419
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009419 100 g 100g
30006512 250 g 250g
30005061 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009419 Supplier Toronto Research Chemicals Supplier No. TRCM110355100G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula Mg . O4 S . 7 H2 O
Molecular Weight (g/mol) 246.47
Synonym Magnesium sulfate heptahydrate, Sulfuric Acid Magnesium Salt (1:1) Heptahydrate, Magnesium Sulfate (MgSO4) Heptahydrate, Magnesium Sulfate Heptahydrate, Magnesium Sulfate Heptahydrate (MgSO4.7H2O), Magnesium Sulfate Hydrate (MgSO4.7H2O);
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg+2].[O-]S(=O)(=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2
Formula Weight 246.0107
Chemical Name or Material Magnesium sulfate heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.