missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate Heptahydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Packaging:
Plastic keg
Quantity:
5 kg
Unit Size:
5kg
Description
5kg, Plastic keg
Specifications
Specifications
| Chemical Name or Material | Magnesium Sulfate Heptahydrate |
| Melting Point | 770°C |
| Name Note | Extra Pure |
| CAS | 10034-99-8 |
| Color | White |
| pH | 5 to 8.2 |
| Physical Form | Solid |
| Quantity | 5 kg |
| Purity Grade Notes | Meets the current analytical specification of BP |
| Molecular Formula | H14MgO11S |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction