missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10697412 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10697412

Brand: Fisher Chemical M/1050/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Magnesium Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 10034-99-8, White, Melting Point: 770 deg.C, Molecular Formula: H14MgO11S, Molecular Weight: 246.466, Packaging: Plastic powder jar, pH: 5 to 8.2

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 770°C
Color White
pH 5 to 8.2
Physical Form Solid
Quantity 500 g
Formula Weight 246.48g/mol
Packaging Plastic powder jar
Vapor Pressure <0.1mmHg at 20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.