missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10743881
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 770.0°C
Color White
pH 5.0 to 9.2 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Assay Percent Range 99% min. (on dry substance) (Complexometry)
Linear Formula MgSO4·7H2O
Merck Index 15, 5755
Solubility Information Solubility in water: 710g/L (20°C). Other solubilities: soluble in glycerol,sparingly soluble in alcohol
IUPAC Name magnesium(2+) heptahydrate sulfate
Formula Weight 246.48
Percent Purity ≥99%
Grade Extra Pure
Loss on Drying 48.0% to 52.0% (450°C)
Packaging Plastic Bottle
Chemical Name or Material Magnesium sulfate heptahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'