missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, ≥99.0%, Reagent Grade, Honeywell™

Product Code. 15617940 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15617940 500 g Poly bottle 500g
15607940 1 kg Poly bottle 1kg
15627940 5 kg Poly bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15617940 Supplier Honeywell Research Chemicals Supplier No. M1880500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1124°C
Color Colorless
pH 5.0 to 9.2 (at 25°C)
Physical Form Solid
Quantity 500 g
Linear Formula MgSO4 · 7H2O
UN Number NONH for all modes of transport
IUPAC Name magnesium;sulfate;heptahydrate
Formula Weight 246.47g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 0.1mmHg (20°C)
Density 1.68g/cu.cm at 20°C; Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium sulfate heptahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.