Learn More
Magnesium sulfate, anhydrous, 99.5% min, Thermo Scientific Chemicals
Use promo code "21741" to get your promotional price.
Description
Magnesium sulfate is suitable for sensitive molecular biology applications. It used as a component of bath salts and beauty product. It is utilized by the athletes to soothe sore muscles and effective in the removal of splinters. It is commonly used as a desiccant in organic synthesis due to its affinity for water. It acts as a brewing salt in beer production to adjust the ion content of the brewing water as well as enhance enzyme action. It plays a vital role as an antidote for barium chloride poisoning. . It is useful in controlling seizures associated with epilepsy, glomerulonephritis, or hypothyroidism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1124°C (decomposition) |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99.5% min |
Solubility Information | Soluble in water. Slightly soluble in alcohol,glycerol. Insoluble in acetone. |
IUPAC Name | magnesium(2+) sulfate |
Formula Weight | 120.37 |
Sensitivity | Hygroscopic |
Density | 2.66 g/mL |
Chemical Name or Material | Magnesium sulfate, anhydrous, anhydrous |
RUO – Research Use Only