missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate, 1M aq. soln.

Product Code. 15426109
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426109 50 mL 50mL
15436109 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426109 Supplier Thermo Scientific Alfa Aesar Supplier No. J63953.AD
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7487-88-9 | MgO4S | 120.36 g/mol

Magnesium sulfate is applied in replacement therapy for hypomagnesemia. It can be used orally as a laxative to relieve occasional constipation, and to treat low levels of magnesium. It works by replacing magnesium in the body and increasing water in the intestines. It is the first-line antiarrhythmic agent for torsades de pointes in cardiac arrest under the 2005 ECC guidelines and for managing quinidine-induced arrhythmias. It is effective in decreasing the risk that pre-eclampsia progresses to eclampsia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium sulfate is applied in replacement therapy for hypomagnesemia. It can be used orally as a laxative to relieve occasional constipation, and to treat low levels of magnesium. It works by replacing magnesium in the body and increasing water in the intestines. It is the first-line antiarrhythmic agent for torsades de pointes in cardiac arrest under the 2005 ECC guidelines and for managing quinidine-induced arrhythmias. It is effective in decreasing the risk that pre-eclampsia progresses to eclampsia.

Solubility
Fully miscible in water. Soluble in alcohol, ether (11.6 mg/ml at 18°C) and glycerin. Insoluble in acetone.

Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium;sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Physical Form Liquid
Quantity 50 mL
Linear Formula MgSO4
Solubility Information Fully miscible in water. Soluble in alcohol,ether (11.6mg/ml at 18°C) and glycerin. Insoluble in acetone.
Formula Weight 120.37
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Magnesium sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.