missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfamate hydrate, 99%

Product Code. 11368158
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368158 250 g 250g
11348158 50 g 50g
11358158 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368158 Supplier Thermo Scientific Alfa Aesar Supplier No. 039381.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13770-91-7 | H4MgN2O6S2 | 216.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13770-91-7
Molecular Formula H4MgN2O6S2
MDL Number MFCD00210632
InChI Key YZVJHCGMTYDKFR-UHFFFAOYSA-L
Synonym magnesium disulfamate, magnesium sulfamate, magnesium disulphamate, sulfamic acid, magnesium salt 2:1, magnesium 2+ ion disulfamate
PubChem CID 166902
IUPAC Name magnesium;disulfamate

Specifications

Quantity 250 g
SMILES NS(=O)(=O)[O-].NS(=O)(=O)[O-].[Mg+2]
Molecular Weight (g/mol) 216.465
Formula Weight 216.48 (Anhydrous)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Magnesium sulfamate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.