missing translation for 'onlineSavingsMsg'
Learn More

Magnesium slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11399428
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399428 10 g 10g
11389428 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11399428 Supplier Thermo Scientific Alfa Aesar Supplier No. 043297.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Quantity 10 g
Odor Odorless
Weight ≈0.09g/slug
Form Slug
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Magnesium slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Annealed
compliance-icons
Product Identifier
  • Magnesium slug
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Self-heating substance Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H228-Flammable solid.
  • H252-Self-heating in large quantities; may catch fire.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use fire-fighting equipment on basis class D to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contain: Magnesium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.