missing translation for 'onlineSavingsMsg'
Learn More

Magnesium silicide, 99.99% (metals basis)

Product Code. 15482327
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482327 250 g 250g
15462327 10 g 10g
15472327 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482327 Supplier Thermo Scientific Alfa Aesar Supplier No. 045518.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Air and Moisture Sensitive; Decomposes in water, HCl | CAS: 22831-39-6 | Mg2Si | 76.695 g/mol

Magnesium silicide is used to produce alloys such as aluminum alloys. It is also used in food, beverages, as a flavor enhancer and as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22831-39-6
Molecular Formula Mg2Si
Molecular Weight (g/mol) 76.695
MDL Number MFCD00016202
InChI Key YTHCQFKNFVSQBC-UHFFFAOYSA-N
Synonym magnesium silicide, dimagnesium silicide, dimagnesium monosilicide, unii-475e6fmg3k, magnesiumsilicid, magnesium, compd. with silicon 2:1, magnesium silicide, powder, magnesium silicide-45 my, magnesium silicide un2624 dangerous when wet
PubChem CID 89858
SMILES [Mg]=[Si]=[Mg]

Specifications

Melting Point 1102°C
Color Blue
Physical Form Pieces
Quantity 250 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Insoluble in water and denser than water.
Formula Weight 76.71
Sensitivity Air and moisture sensitive
Density 2
Chemical Name or Material Magnesium silicide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.