missing translation for 'onlineSavingsMsg'
Learn More

Magnesium salicylate tetrahydrate, tech.

Product Code. 11318158
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318158 500 g 500g
11308158 100 g 100g
11398148 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11318158 Supplier Thermo Scientific Alfa Aesar Supplier No. 039379.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18917-95-8 | C14H18MgO10 | 370.59 g/mol

It is used to treat mild to moderate muscular pain. It is also used to treat headaches, general back pain, and certain joint pains like arthritis. It is found in a variety of over-the-counter (OTC) medications as an anti-inflammatory, primarily for back-pain relief.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18917-95-8
Molecular Formula C14H18MgO10
Molecular Weight (g/mol) 370.59
MDL Number MFCD01766176,MFCD00045815
InChI Key NBQBEWAYWAMLJJ-UHFFFAOYSA-L
Synonym magnesium salicylate
PubChem CID 131674114
IUPAC Name magnesium;2-hydroxybenzoic acid;2-oxidobenzoate;hydrate
SMILES O.O.O.O.[Mg++].OC1=CC=CC=C1C([O-])=O.OC1=CC=CC=C1C([O-])=O

Specifications

Odor Odorless
Quantity 500 g
Sensitivity Air Sensitive
Merck Index 14,5672
Solubility Information Soluble in water and alcohol
Formula Weight 370.60 (298.60 Anhydrous)
Grade Technical
Physical Form Powder
Chemical Name or Material Magnesium salicylate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.