missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium salicylate tetrahydrate, tech., Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
It is used to treat mild to moderate muscular pain. It is also used to treat headaches, general back pain, and certain joint pains like arthritis. It is found in a variety of over-the-counter (OTC) medications as an anti-inflammatory, primarily for back-pain relief.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Odor | Odorless |
Quantity | 2.5 kg |
Sensitivity | Air Sensitive |
Merck Index | 14,5672 |
Solubility Information | Soluble in water and alcohol |
Formula Weight | 370.60 (298.60 Anhydrous) |
Grade | Technical |
Physical Form | Powder |
Chemical Name or Material | Magnesium salicylate tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'