missing translation for 'onlineSavingsMsg'
Learn More

Magnesium rod, 3.3cm (1.3in) dia x 30cm (12in) long, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11375759
Change view
Click to view available options
Quantity:
1 Pc.
25 Pc.
5 Pc.
Unit Size:
Each
Pack of 25
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375759 25 Pc. Pack of 25
11365759 1 Pc. Each
15485925 5 Pc. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11375759 Supplier Thermo Scientific Alfa Aesar Supplier No. 010231.KM
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Quantity 25 Pc.
Odor Odorless
Weight ≈450g/pc
Form Rod
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Magnesium rod, 3.3cm (1.3 in.) dia. x 30cm (12 in.) long

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.