missing translation for 'onlineSavingsMsg'
Learn More

Magnesium pyrophosphate

Product Code. 11363728
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363728 250 g 250g
11383728 50 g 50g
11373728 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363728 Supplier Thermo Scientific Alfa Aesar Supplier No. 039317.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-24-7 | Mg2O7P2 | 222.551 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-24-7
Molecular Formula Mg2O7P2
Molecular Weight (g/mol) 222.551
MDL Number MFCD00049482
InChI Key XZTWHWHGBBCSMX-UHFFFAOYSA-J
Synonym magnesium pyrophosphate, unii-al34y660jv, diphosphoric acid, magnesium salt 1:2, dimagnesium diphosphate, acmc-1but0, 2mg.p2o7, magnesium pyrophosphate 50g, dimagnesium phosphonato phosphate, dimagnesium 2+ diphosphate, dimagnesium 2+ ion diphosphate
PubChem CID 122213
IUPAC Name dimagnesium;phosphonato phosphate
SMILES [O-]P(=O)([O-])OP(=O)([O-])[O-].[Mg+2].[Mg+2]

Specifications

Color White
Physical Form Powder
Quantity 250 g
Merck Index 14,5684
Solubility Information Soluble in dilute mineral acids. Insoluble in water.
Formula Weight 222.57
Chemical Name or Material Magnesium pyrophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.