missing translation for 'onlineSavingsMsg'
Learn More

Magnesium powder, Honeywell™

Product Code. 16676263 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
25 kg
Unit Size:
100g
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16676263 100 g 100g
16696263 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16676263 Supplier Honeywell Research Chemicals Supplier No. 13112H100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, ≥99%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
PubChem CID 5462224
ChEBI CHEBI:25107

Specifications

Appearance Complying
Quantity 100 g
Packaging Plastic Bottle
Form Powder
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.