missing translation for 'onlineSavingsMsg'
Learn More

Magnesium phthalocyanine, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7050556
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1661-03-6
C32H18MgN8O
554.856
MFCD00041953
HTNHYCHFBSVBID-UHFFFAOYSA-N
magnesium phthalocyanine
91872988
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O.[Mg+2]
This item is not returnable. View return policy
1661-03-6
C32H18MgN8O
554.856
MFCD00041953
HTNHYCHFBSVBID-UHFFFAOYSA-N
magnesium phthalocyanine
91872988
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O.[Mg+2]

Magnesium phthalocyanine is used in preparing membrane sensors for cyanide ions. Also used in preparation of membrane sensors for cyanide ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Soluble in concentrated sulfuric acid.
Formula Weight 536.84
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material Magnesium phthalocyanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'