missing translation for 'onlineSavingsMsg'
Learn More

Magnesium phosphate pentahydrate, P{2}O{5} 33%

Product Code. 11393427
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393427 5 kg 5kg
11303437 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393427 Supplier Thermo Scientific Alfa Aesar Supplier No. 018219.A7
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10233-87-1 | H10Mg3O13P2 | 352.93 g/mol

Magnesium phosphate pentahydrate is used in the preparation of magnesium calcium phosphate biocement(MCPB) for bone regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10233-87-1
Molecular Formula H10Mg3O13P2
Molecular Weight (g/mol) 352.93
MDL Number MFCD00150252
InChI Key QZYXGKYJDBUYES-UHFFFAOYSA-H
Synonym magnesium phosphate pentahydrate, magnesium phosphate usp, phosphoric acid, magnesium salt 2:3 , pentahydrate, 3mg.2po4.5h2o, trimagnesium phosphate pentahydrate, magnesium phosphate 3:2 pentahydrate, magnesium phosphate, tribasic pentahydrate, magnesium phosphate pentahydrate, p2o, trimagnesium 2+ pentahydrate diphosphate
PubChem CID 23724945
IUPAC Name trimagnesium;diphosphate;pentahydrate
SMILES O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Mg+2].[Mg+2].[Mg+2]

Specifications

Color White
Physical Form Crystals
Quantity 5 kg
Appearance White crystals
Assay Percent Range P2O5 33%
Solubility Information Insoluble in water.
Formula Weight 352.93 (262.86 Anhydrous)
Chemical Name or Material Magnesium phosphate pentahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.